Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mppb-1
Entrez Gene ID
177725
Gene Name
Mitochondrial-processing peptidase subunit beta
Gene Aliases
CELE_ZC410.2
Chromosome Location
Chr.IV: 9074575 - 9076574 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069175.8 NP_501576.2 6 - 7 813 69

View Full Product Details