Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mrpl-42
- Entrez Gene ID
-
36805074
- Gene Name
- Mitochondrial Ribosomal Protein, Large
- Gene Aliases
-
CELE_ZC410.10
- Chromosome Location
-
Chr.IV: 9092217 - 9094456 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069179.9 |
NP_501580.3 |
5 - 6 |
904 |
96 |
- Gene Symbol
- lpl-1
- Entrez Gene ID
-
177729
- Gene Name
- Putative lipoyltransferase 2, mitochondrial
- Gene Aliases
-
CELE_ZC410.7
- Chromosome Location
-
Chr.IV: 9092173 - 9094479 on Build WBcel235
View Full Product Details