Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
del-2
Entrez Gene ID
177796
Gene Name
protein-coding
Gene Aliases
CELE_F58G6.6
Chromosome Location
Chr.IV: 9636905 - 9642025 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268482.3 NP_001255411.1 1 - 2 97 94

View Full Product Details