Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gln-6
Entrez Gene ID
177808
Gene Name
Gln-synt_C domain-containing protein
Gene Aliases
CELE_C28D4.3
Chromosome Location
Chr.IV: 9739261 - 9741732 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069332.5 NP_501733.1 3 - 4 786 63

View Full Product Details