Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-97
Entrez Gene ID
177810
Gene Name
protein-coding
Gene Aliases
CELE_H27C11.1
Chromosome Location
Chr.IV: 9757646 - 9764497 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047489.4 NP_001040954.1 2 - 3 406 60
GenBank mRNA AF332211.1 - 2 - 3 406 60

View Full Product Details