Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sna-2
Entrez Gene ID
177816
Gene Name
Small Nuclear RNA (snRNA) Associated protein
Gene Aliases
CELE_T13F2.7
Chromosome Location
Chr.IV: 9774477 - 9777634 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069343.6 NP_501744.1 6 - 7 1915 72

View Full Product Details