Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npp-1
Entrez Gene ID
177836
Gene Name
protein-coding
Gene Aliases
CELE_K07F5.13
Chromosome Location
Chr.IV: 9858488 - 9864031 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069370.4 NP_501771.1 3 - 4 312 57

View Full Product Details