Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-3
Entrez Gene ID
177883
Gene Name
Glutathione S-transferase 3
Gene Aliases
CELE_K08F4.11
Chromosome Location
Chr.IV: 10139380 - 10141539 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069445.7 NP_501846.1 3 - 4 598 102
GenBank mRNA AF010241.1 - 1 - 2 128 102

View Full Product Details