Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-7
Entrez Gene ID
177931
Gene Name
Guanine nucleotide-binding protein alpha-7 subunit
Gene Aliases
CELE_R10H10.5
Chromosome Location
Chr.IV: 10404008 - 10408747 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069520.3 NP_501921.1 3 - 4 471 85
GenBank mRNA AY008130.1 - 3 - 4 460 85

View Full Product Details