Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ced-5
Entrez Gene ID
177942
Gene Name
Uncharacterized protein
Gene Aliases
CELE_C02F4.1
Chromosome Location
Chr.IV: 10484110 - 10492952 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069537.7 NP_501938.1 4 - 5 732 126
GenBank mRNA AF038576.1 - 4 - 5 753 126

View Full Product Details