Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zim-2
Entrez Gene ID
177948
Gene Name
C2H2-type domain-containing protein
Gene Aliases
CELE_T07G12.10
Chromosome Location
Chr.IV: 10547086 - 10558218 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383192.1 7 - 8 2568 136

View Full Product Details