Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
epi-1
Entrez Gene ID
177956
Gene Name
protein-coding
Gene Aliases
CELE_K08C7.3
Chromosome Location
Chr.IV: 10666821 - 10679291 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028110.4 NP_001023281.1 12 - 13 9970 80
NM_001307078.4 NP_001294007.1 12 - 13 9941 80
NM_001307079.3 NP_001294008.1 13 - 14 9991 80
GenBank mRNA AB001074.1 - 12 - 13 9964 80

View Full Product Details