Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ugt-54
- Entrez Gene ID
-
188885
- Gene Name
- UDP-GlucuronosylTransferase
- Gene Aliases
-
CELE_T25B9.7
- Chromosome Location
-
Chr.IV: 10761698 - 10764496 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069595.6 |
NP_501996.2 |
1 - 2 |
173 |
146 |
- Gene Symbol
- T25B9.11
- Entrez Gene ID
-
4927051
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T25B9.11
- Chromosome Location
-
Chr.IV: 10761622 - 10765407 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383197.1 |
|
1 - 2 |
249 |
146 |
View Full Product Details