Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpt-2
Entrez Gene ID
178025
Gene Name
protein-coding
Gene Aliases
CELE_R07H5.2
Chromosome Location
Chr.IV: 11179645 - 11183001 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047512.4 NP_001040977.2 1 - 2 164 66

View Full Product Details