Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prx-14
Entrez Gene ID
178026
Gene Name
Pex14_N domain-containing protein
Gene Aliases
CELE_R07H5.1
Chromosome Location
Chr.IV: 11183132 - 11184310 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069696.5 NP_502097.1 1 - 2 267 85

View Full Product Details