Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-7
Entrez Gene ID
178035
Gene Name
Nuclear hormone receptor family member nhr-7
Gene Aliases
CELE_F54D1.4
Chromosome Location
Chr.IV: 11266265 - 11271362 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069716.7 NP_502117.2 4 - 5 559 112
NR_138498.1 4 - 5 549 112
GenBank mRNA AF083225.1 - 3 - 4 1356 112

View Full Product Details