Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpm-1
Entrez Gene ID
176874
Gene Name
protein-coding
Gene Aliases
CELE_Y66H1A.2
Chromosome Location
Chr.IV: 445677 - 449837 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067530.6 NP_499931.2 2 - 3 570 94

View Full Product Details