Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
efn-4
Entrez Gene ID
176882
Gene Name
Ephrin-4
Gene Aliases
CELE_F56A11.3
Chromosome Location
Chr.IV: 559234 - 568148 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067546.5 NP_499947.1 3 - 4 959 139
GenBank mRNA AF410936.1 - 3 - 4 955 139

View Full Product Details