Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-40
Entrez Gene ID
176900
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_F56B3.10
Chromosome Location
Chr.IV: 760502 - 762419 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067580.5 NP_499981.1 3 - 4 493 74

View Full Product Details