Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-52
Entrez Gene ID
176904
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_F56B3.7
Chromosome Location
Chr.IV: 778784 - 784182 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067587.2 NP_499988.1 3 - 4 383 142

View Full Product Details