Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-30
Entrez Gene ID
186551
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_F58H7.3
Chromosome Location
Chr.IV: 924882 - 939736 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067614.4 NP_500015.3 6 - 7 768 81

View Full Product Details