Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atg-3
Entrez Gene ID
176921
Gene Name
protein-coding
Gene Aliases
CELE_Y55F3AM.4
Chromosome Location
Chr.IV: 1010488 - 1012502 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067623.6 NP_500024.1 1 - 2 318 81

View Full Product Details