Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
csn-4
Entrez Gene ID
176927
Gene Name
COP9 signalosome complex subunit 4
Gene Aliases
CELE_Y55F3AM.15
Chromosome Location
Chr.IV: 1077462 - 1087013 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067633.6 NP_500034.1 4 - 5 630 87

View Full Product Details