Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pbs-1
Entrez Gene ID
176978
Gene Name
Proteasome subunit beta
Gene Aliases
CELE_K08D12.1
Chromosome Location
Chr.IV: 1708803 - 1709745 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067724.10 NP_500125.2 1 - 2 82 63

View Full Product Details