Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cng-2
Entrez Gene ID
190732
Gene Name
protein-coding
Gene Aliases
CELE_Y76B12C.1
Chromosome Location
Chr.IV: 2037191 - 2043549 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356909.3 NP_001343690.1 3 - 4 520 57

View Full Product Details