The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001028280.5 | NP_001023451.1 | 5 - 6 | 1307 | 97 |
GenBank mRNA | AB009567.1 | - | 4 - 5 | 630 | 97 |