Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ric-8
Entrez Gene ID
177048
Gene Name
protein-coding
Gene Aliases
CELE_Y69A2AR.2
Chromosome Location
Chr.IV: 2642977 - 2650111 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028390.5 NP_001023561.1 1 - 2 149 84
NM_001307677.3 NP_001294606.1 1 - 2 141 84
NM_001383056.1 1 - 2 149 84
NM_001383057.1 1 - 2 151 84
GenBank mRNA AF288812.1 - 1 - 2 149 84

View Full Product Details