Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-70
Entrez Gene ID
182472
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_C09G12.6
Chromosome Location
Chr.IV: 3477392 - 3479011 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067954.2 NP_500355.2 2 - 3 269 156

View Full Product Details