Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smgl-2
Entrez Gene ID
177135
Gene Name
protein-coding
Gene Aliases
CELE_Y37E11AM.1
Chromosome Location
Chr.IV: 3726164 - 3735496 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307445.3 NP_001294374.1 3 - 4 476 62

View Full Product Details