Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-19
Entrez Gene ID
177150
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_F56D6.4
Chromosome Location
Chr.IV: 3919806 - 3921613 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068035.2 NP_500436.2 3 - 4 593 148

View Full Product Details