Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-72
Entrez Gene ID
177152
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_Y46C8AL.5
Chromosome Location
Chr.IV: 3944816 - 3947364 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068041.6 NP_500442.3 3 - 4 674 74

View Full Product Details