Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-20
Entrez Gene ID
177189
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_F36A4.8
Chromosome Location
Chr.IV: 4246734 - 4247486 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068121.7 NP_500522.1 2 - 3 256 70

View Full Product Details