Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ehn-3
Entrez Gene ID
191363
Gene Name
protein-coding
Gene Aliases
CELE_ZK616.10
Chromosome Location
Chr.IV: 5416699 - 5419222 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028444.3 NP_001023615.1 5 - 6 648 80
NM_001028445.1 NP_001023616.1 3 - 4 489 80
NM_001028446.1 NP_001023617.1 2 - 3 360 80

View Full Product Details