Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-24
Entrez Gene ID
177380
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_C49A9.8
Chromosome Location
Chr.IV: 6206821 - 6209459 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068530.6 NP_500931.3 7 - 8 1181 78

View Full Product Details