Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-18
Entrez Gene ID
189745
Gene Name
protein-coding
Gene Aliases
CELE_Y39C12A.3
Chromosome Location
Chr.IV: 12227717 - 12229779 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307201.4 NP_001294130.1 5 - 6 630 98
NR_131614.1 4 - 5 472 98

View Full Product Details