Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-20
Entrez Gene ID
188045
Gene Name
protein-coding
Gene Aliases
CELE_T04A11.7
Chromosome Location
Chr.IV: 12495496 - 12497923 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356933.3 NP_001343588.1 - 1436 168
NM_001356934.1 NP_001343704.1 3 - 4 340 168

View Full Product Details