Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vha-7
Entrez Gene ID
178219
Gene Name
protein-coding
Gene Aliases
CELE_C26H9A.1
Chromosome Location
Chr.IV: 12640462 - 12650653 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383222.1 15 - 16 2108 138
NM_070018.6 NP_502419.2 10 - 11 1376 138
GenBank mRNA AB055112.1 - 12 - 13 1773 138

View Full Product Details