Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-6
Entrez Gene ID
178231
Gene Name
Innexin-6
Gene Aliases
CELE_C36H8.2
Chromosome Location
Chr.IV: 12746584 - 12749721 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070034.5 NP_502435.1 4 - 5 438 117

View Full Product Details