Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-1
Entrez Gene ID
186053
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_F49E11.9
Chromosome Location
Chr.IV: 13056326 - 13057351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070101.3 NP_502502.1 3 - 4 590 91

View Full Product Details