Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srt-49
Entrez Gene ID
189834
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_Y41E3.14
Chromosome Location
Chr.IV: 15092813 - 15098394 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070421.3 NP_502822.2 5 - 6 734 137

View Full Product Details