Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-15
Entrez Gene ID
190889
Gene Name
protein-coding
Gene Aliases
CELE_Y105C5B.6
Chromosome Location
Chr.IV: 15906582 - 15910091 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047546.3 NP_001041011.1 6 - 7 764 100
NM_001047547.3 NP_001041012.1 4 - 5 404 100
NM_001307189.3 NP_001294118.1 6 - 7 782 100
NM_001307190.3 NP_001294119.1 4 - 5 422 100

View Full Product Details