Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-11
Entrez Gene ID
186779
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_H25K10.3
Chromosome Location
Chr.IV: 16219599 - 16223232 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070521.2 NP_502922.1 2 - 3 485 117

View Full Product Details