Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fipr-28
Entrez Gene ID
178462
Gene Name
FIP (Fungus-Induced Protein) Related
Gene Aliases
CELE_Y51H4A.26
Chromosome Location
Chr.IV: 16627976 - 16628444 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070567.7 NP_502968.1 2 - 2 309 82

View Full Product Details