Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-82
Entrez Gene ID
259811
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_W03F9.6
Chromosome Location
Chr.V: 145094 - 146952 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070743.2 NP_503144.1 1 - 2 316 70

View Full Product Details