Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cogc-2
Entrez Gene ID
177473
Gene Name
Conserved oligomeric Golgi complex subunit 2
Gene Aliases
CELE_C06G3.10
Chromosome Location
Chr.IV: 7036867 - 7039580 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068691.7 NP_501092.1 3 - 4 528 86
GenBank mRNA Z34976.1 - - 526 86

View Full Product Details