Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-27
Entrez Gene ID
191204
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_ZK6.3
Chromosome Location
Chr.V: 400021 - 401602 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070788.4 NP_503189.2 1 - 2 413 60

View Full Product Details