Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-33
Entrez Gene ID
191852
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_M02H5.9
Chromosome Location
Chr.V: 441820 - 443293 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070802.1 NP_503203.1 3 - 4 514 69

View Full Product Details