Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-31
Entrez Gene ID
187407
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_M02H5.2
Chromosome Location
Chr.V: 472889 - 474631 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070812.1 NP_503213.1 3 - 4 933 115

View Full Product Details