Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-25
Entrez Gene ID
185172
Gene Name
protein-coding
Gene Aliases
CELE_F31F4.14
Chromosome Location
Chr.V: 672245 - 674313 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313162.1 NP_001300091.1 3 - 4 399 100
NM_070888.2 NP_503289.2 3 - 4 594 100

View Full Product Details