Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-5
Entrez Gene ID
190063
Gene Name
protein-coding
Gene Aliases
CELE_Y49C4A.4
Chromosome Location
Chr.V: 700079 - 701669 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070898.1 NP_503299.1 2 - 3 317 120

View Full Product Details